Learn More
anti-PRKAG2, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00051422-PW1.1Set
Product Code. 16014559
  • 843.00€ / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-PRKAG2 (300μL), Antibody pair for WB: mouse polyclonal anti-PRKAG2 (50μL)
Specifications
PRKAG2
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
AAKG/AAKG2/CMH6/H91620p/WPWS
Mouse, Rabbit
RUO
51422
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
PRKAG2
PRKAG2
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-PRKAG2 (300 ul) 2. Antibody pair for WB: mouse polyclonal anti-PRKAG2 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.