missing translation for 'onlineSavingsMsg'
Learn More

anti-PPP2R3B Ab-2 Matched Antibody Pair, Abnova™

Product Code. 16183334
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16183334 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16183334 Supplier Abnova Supplier No. H00028227AP12.1Set
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PPP2R3B (100μg); Detection antibody: mouse monoclonal anti-PPP2R3B, IgG2b Kappa (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PPP2R3B

Host Species: Rabbit, Mouse
Isotype: IgG, IgG2b κ
Species Reactivity: Hu

ELISA Pair

Specifications

Antigen PPP2R3B
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PPP2R3B.
Gene PPP2R3B
Gene Alias NY-REN-8/PPP2R3L/PPP2R3LY/PR48
Gene Symbols PPP2R3B
Host Species Mouse, Rabbit
Purification Method Affinity Purified
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28227
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PPP2R3B (100 μg) 2. Detection antibody: mouse monoclonal anti-PPP2R3B, IgG2b Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Isotype IgG, IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.