missing translation for 'onlineSavingsMsg'
Learn More

p21 Mouse anti-Human, Unlabeled, Clone: 6B6, BD

Product Code. 15897618
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15897618 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15897618 Supplier BD Biosciences Supplier No. 554228

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

p21 belongs to a class of tumor suppressors including p16 and p27 which control progression through the cell cycle by inhibiting the activity of cyclin-cdk complexes. p21 is also known as senescent cell-derived inhibitor 1 (Sdi1), wild-type p53-activated fragment 1 (Waf1), Cdk-interacting protein 1 (Cip1), p21 and p53-regulated inhibitor of Cdks (Pic1). p21 mRNA is expressed at higher levels in senescent fibroblasts than in actively growing cells. When introduced into proliferating foreskin fibroblasts, p21 inhibits DNA synthesis and cell cycle arrest. p53 can activate p21 gene expression via a p53 binding site identified in the promoter of the p21 gene.

Host Species: Mouse
Clone: 6B6
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human p21
Formula Weight [Chemical]: 21kDa

Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p21
Applications Western Blot
Classification Monoclonal
Clone 6B6
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human p21
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.