Learn More
anti-NRP1, Human, Matched Antibody Pair, Abnova™
Product Code. 16192894
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16192894 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16192894 Supplier Abnova Supplier No. H00008829AP62.1Set
 Restricted Item
Estimated Shipment: 06-10-2026
Includes
Capture antibody: mouse monoclonal anti-NRP1 (100μg) , Detection antibody: rabbit MaxPab affinity purified polyclonal anti-NRP1 (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Specifications
Antigen NRP1
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NRP1.
Gene NRP1
Gene Alias BDCA4/CD304/DKFZp686A03134/DKFZp781F1414/NP1/NRP/VEGF165R
Gene Symbols NRP1
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8829
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-NRP1 (100 μg) 2. Detection antibody: rabbit MaxPab™ affinity purified polyclonal anti-NRP1 (50 μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid