Learn More
nuclear factor (erythroid-derived 2)-like 2, Mouse, Clone: 3G7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant NFE2L2.
Brand: Abnova H00004780-M04.100ug
Description
NFE2 (MIM 601490), NFE2L1 (MIM 163260), and NFE2L2 comprise a family of human genes encoding basic leucine zipper (bZIP) transcription factors. They share highly conserved regions that are distinct from other bZIP families, such as JUN (MIM 165160) and FOS (MIM 164810), although remaining regions have diverged considerably from each other (Chan et al., 1995 [PubMed 7868116]).[supplied by OMIM
Sequence: SPSVASPEHSVESSSYGDTLLGLSDSEVEELDSAPGSVKQNGPKTPVHSSGDMVQPLSPSQGQSTHVHDAQCENTPEKELPVSPGHRKTPFTKDKHSSRLSpecifications
| nuclear factor (erythroid-derived 2)-like 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_006164 | |
| NFE2L2 | |
| NFE2L2 (NP_006155.2, 351 a.a. ∼ 450 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3G7 | |
| Mouse monoclonal antibody raised against a full length recombinant NFE2L2. | |
| NFE2L2 | |
| NRF2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 4780 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.