Learn More
anti-NEDD9, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00004739-PW1.1Set
Product Code. 16182364
  • 843.00€ / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-NEDD9 (300μL), Antibody pair for WB: mouse purified polyclonal anti-NEDD9 (50μg)
Specifications
NEDD9
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
CAS-L/CAS2/CASL/CASS2/HEF1/dJ49G10.2/dJ761I2.1
Mouse, Rabbit
RUO
4739
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
NEDD9
NEDD9
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-NEDD9 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-NEDD9 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.