missing translation for 'onlineSavingsMsg'
Learn More

Anti-NCX1 antibody [C2C12], Abcam

Product Code. 32047559 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32047559 20 μg 20µg
32045676 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32047559 Supplier Abcam Supplier No. ab286920ug
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse Monoclonal NCX1 antibody. Suitable for Flow Cyt (Intra), IHC-P, IHC-Fr and reacts with Human samples. Cited in 34 publications.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen SLC8A1 (NCX1)
Applications Flow Cytometry (Intracellular Staining), Immunohistochemistry (FFPE), Immunohistochemistry (Frozen)
Cellular Localization Cell membrane; Multi-pass membrane protein
Classification Monoclonal
Clone C2C12
Conjugate Unconjugated
Formulation Preservative: 0.02% Sodium azide, Constituents: PBS, 6.97% L-Arginine
Gene SLC8A1
Gene Accession No. P32418
Host Species Mouse
Immunogen SLC8A1 (NCX1)
Quantity 20 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgM
Shipping Condition Blue Ice
Target Gene SLC8A1
Target Molecule SLC8A1 (NCX1)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.