missing translation for 'onlineSavingsMsg'
Learn More

NADK, Human, Matched Antibody Pair (AP11), Abnova™

Product Code. 16103654
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16103654 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16103654 Supplier Abnova Supplier No. H00065220AP11.1Set

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-NADK (100μg); Detection antibody: mouse monoclonal anti-NADK, IgG2a Kappa (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NADK.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

ELISA Pair (Recombinant Protein)

Specifications

Antigen NADK
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NADK.
Gene NADK
Gene Alias FLJ13052/FLJ37724/dJ283E3.1
Gene Symbols NADK
Host Species Mouse, Rabbit
Purification Method Affinity Purified
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65220
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-NADK (100 μg) 2. Detection antibody: mouse monoclonal anti-NADK, IgG2a Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.