Learn More
Na+, K+ ATPase β3 Mouse anti-Human, Unlabeled, Clone: 46, BD
Mouse Monoclonal Antibody
Specifications
| Antigen | Na+, K+ ATPase β3 |
|---|---|
| Clone | 46 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Na+,K+-ATPase is an integral plasma membrane protein that maintains the intracellular concentrations of Na+ and K+ ions. It is a P class pump that is composed of two α and two β transmembrane subunits. It is also thought to contain a γ subunit. The 120kDa α subunit represents the catalytic site. Although the 50kDa β subunit mediates proper folding of newly synthesized α subunits, it is not directly involved in ion
transfer. There are four known α isoforms (α1, α2, α3, and α4) and three β isoforms (β1, β2, and β3). The Na+,K+-ATPase is ubiquitously expressed in the plasma membrane. However, the individual subunits have different tissue distributions: α1 is ubiquitous; α2 is in skeletal muscle, brain, and heart; α3 is in brain and heart; α4 is present in testis and skeletal muscle; β1 is in most tissues; and β2 is primarily in neural tissues. Different combinations of the α and β subunits can alter the kinetics of enzyme activity. The Na+,K+-ATPase transfers three Na+ ions out of, and two K+ ions into, the cell upon hydrolysis of one ATP molecule. Maintenance of intracellular Na+,K+ levels is necessary for volume regulation, action potentials, and secondary active transport.
Immunofluorescence, Western Blotting
Specifications
| Na+, K+ ATPase β3 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 46 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Human Na+, K+ ATPase β3 aa. 124-243 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.