Learn More
anti-MTDH, Human, IP-WB Antibody Pair, Abnova™
Description
Includes
Antibody pair for IP: rabbit polyclonal anti-MTDH (300μL), Antibody pair for WB: mouse purified polyclonal anti-MTDH (50μg)
Specifications
Specifications
| Antigen | MTDH |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | MTDH |
| Gene Alias | 3D3/AEG-1/AEG1/LYRIC |
| Gene Symbols | MTDH |
| Host Species | Mouse, Rabbit |
| Quantity | 1 Set |
| Show More |