Learn More
mitochondrial ribosomal protein S23, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MRPS23 protein.
Brand: Abnova H00051649-B01P.50ug
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. A pseudogene corresponding to this gene is found on chromosome 7p. [provided by RefSeq
Sequence: MAGSRLETVGSIFSRTRDLVRAGVLKEKPLWFDVYDAFPPLRGPVFQRPRVRYGKAKAPIQDIWYHEDRIRAKFYSVYGSGQRAFDLFNPNFKSTCQRFVEKYTELQKLGETDEEKLFVETGKALLAEGVILRRVGEARTQHGGSHVSRKSEHLSVRPQTALEENETQKEVPQDQHLEAPADQSKGLLPPSpecifications
| mitochondrial ribosomal protein S23 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human MRPS23 protein. | |
| MRPS23 | |
| CGI-138/HSPC329/MRP-S23 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 51649 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC000242.1 | |
| MRPS23 | |
| MRPS23 (AAH00242.1, 1 a.a. ∼ 190 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.