Learn More
Anti-Mouse PD-1/CD279 (RMP1-30), Proteintech
Rat Monoclonal Antibody
Brand: Proteintech 65142-1-IG-500UG
This item is not returnable.
View return policy
Description
The PD-1/CD279 antibody from Proteintech is a rat monoclonal antibody generated with mouse pd-1 transfected bhk cells of mouse PD-1/CD279. This antibody recognizes mouse antigen. The PD-1/CD279 antibody has been validated for the following applications: FC analysis.Specifications
| PD-1/CD279 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Protein A purification | |
| RUO | |
| 18566 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| RMP1-30 | |
| Unconjugated | |
| Mouse PD-1 transfected BHK cells | |
| 500 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG2b κ |