missing translation for 'onlineSavingsMsg'
Learn More

Ig Goat anti-Mouse, Brilliant Violet 480, Polyclonal, BD

Product Code. 15873939
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15873939 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15873939 Supplier BD Biosciences Supplier No. 564877
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Goat Polyclonal Antibody

BD Horizon™ BV480 Goat Anti-Mouse Ig is intended to be a second-step reagent for immunofluorescent staining of cells pre-stained with Mouse Ig primary antibodies. It is reactive with the heavy and light chains of mouse primary antibodies having the IgG1, IgG2a, IgG2b, IgG3, IgM, and IgA isotypes. Minimal background staining of human and rat cells occurs in the absence of a primary mouse antibody. In addition, the BD Horizon BV480 Goat Anti-Mouse Ig stains mouse B lymphocytes with little non-specific staining of other cells. Therefore, it is useful as a primary immunofluorescent reagent for staining mouse B cells and antibody-producing cells and as a secondary reagent for staining human and rat cells pre-stained with mouse Ig primary antibodies.

The antibody was conjugated to BD Horizon BV480 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye has been exclusively developed by BD Biosciences as an optimal dye for imaging applications. BD Horizon BV480 has an Ex Max at 436nm and Em Max at 477 nm. The use of a mounting reagent (eg, ProLong™ Gold) is highly recommended to maximize the photostability of BV480.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig
Applications Immunofluorescence
Classification Polyclonal
Clone Polyclonal
Concentration 0.2mg/mL
Conjugate Brilliant Violet 480
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Goat
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype Ig
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.