Learn More
Anti-Mouse Foxp3 (3G3), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65089-1-IG-100UG
This item is not returnable.
View return policy
Description
The Foxp3 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of mouse Foxp3. This antibody recognizes mouse antigen. The Foxp3 antibody has been validated for the following applications: IHC, FC (Intra) analysis.Specifications
| Foxp3 | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| 100 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG1 κ |
| Immunohistochemistry, Flow Cytometry (Intracellular Staining) | |
| 3G3 | |
| Unconjugated | |
| Affinity purification | |
| RUO | |
| 20371 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |