missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse CD49b (DX5), Proteintech
Rat Monoclonal Antibody
Brand: Proteintech 65065-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD49b antibody from Proteintech is a rat monoclonal antibody generated with nk cells isolated from c57bl/6 mice of mouse CD49b. This antibody recognizes mouse antigen. The CD49b antibody has been validated for the following applications: FC analysis.Specifications
| CD49b | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Affinity purification | |
| RUO | |
| 16398 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| DX5 | |
| Unconjugated | |
| NK cells isolated from C57BL/6 mice | |
| 500 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction