missing translation for 'onlineSavingsMsg'
Learn More

CD36 Mouse anti-Mouse, PE, Clone: CRF D-2712, BD

Product Code. 15892159
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15892159 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15892159 Supplier BD Biosciences Supplier No. 562702
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CRF D-2712 monoclonal antibody specifically binds to mouse and rat CD36, an 88-kDa glycoprotein member of the class B scavenger receptor family. CD36 is a multifunctional receptor that has a variety of ligands and is expressed on many cell types. Consequently, it is known by several names, including macrophage scavenger receptor, platelet glycoprotein IV, and fatty acid translocase. Cells that express CD36 include platelets, megakaryocytes, monocytes/macrophages, dendritic cells, erythroid precursors, microvascular endothelia, skeletal, cardiac, and smooth muscle cells, adipocytes, and epithelia of retina, breast, and intestine. This multi-ligand receptor functions in the clearance of apoptotic cells and abnormal erythrocytes (in malaria and sickle-cell anemia), the transport of long-chain fatty acids and oxidized low-density lipoproteins, the regulation of angiogenesis by mediating the effects of thrombospondin-1, and the mediation of coronary vasoconstriction. Crosslinking of cell-surface CD36 with plate-bound CRF D-2712 antibody promotes internalization of photoreceptor outer segment fragments by cultured retinal pigment epithelium.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD36
Applications Flow Cytometry
Classification Monoclonal
Clone CRF D-2712
Concentration 0.2mg/mL
Conjugate PE
Description Platelet glycoprotein 4; PAS-4; GPIIIB; Fatty acid translocase; Scavenger R
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant mouse CD36
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgA κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.