missing translation for 'onlineSavingsMsg'
Learn More

Anti-Mouse CD134/OX40 (OX-86), Proteintech

Rat Monoclonal Antibody

57.00€ - 171.00€

Specifications

Antigen CD134/OX40
Clone OX-86
Concentration 500 μg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30233966
View Documents
Proteintech
65136-1-IG-100UG
100 μg
57.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30236889
View Documents
Proteintech
65136-1-IG-500UG
500 μg
171.00€
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The CD134/OX40 antibody from Proteintech is a rat monoclonal antibody generated with n/a of mouse CD134/OX40. This antibody recognizes mouse antigen. The CD134/OX40 antibody has been validated for the following applications: FC analysis.
Specifications

Specifications

CD134/OX40
500 μg/mL
Monoclonal
Liquid
RUO
22163
Primary
Store at 2-8°C. Stable for one year after shipment.
OX-86
Flow Cytometry
Unconjugated
Rat
Mouse
IgG1 κ
Affinity purification
Flow Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.