missing translation for 'onlineSavingsMsg'
Learn More

makorin ring finger protein 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Product Code. 16158788
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16158788 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16158788 Supplier Abnova Supplier No. H00023609D01P.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against a full-length human MKRN2 protein.

Members of the makorin family, including MKRN2, have a characteristic zinc finger composition that suggests that they are ribonucleoproteins (Gray et al., 2001 [PubMed 11597136]).[supplied by OMIM

Sequence: MSTKQITCRYFMHGVCREGSQCLFSHDLANSKPSTICKYYQKGYCAYGTRCRYDHTRPSAAAGGAVGTMAHSVPSPAFHSPHPPSEVTASIVKTNSHEPGKREKRTLVLRDRNLSGMAERKTQPSMVSNPGSCSDPQPSPEMKPHSYLDAIRSGLDDVEASSSYSNEQQLCPYAAAGECRFGDACVYLHGEVCEICRLQVLHPFDPEQRKAHEKICMLTFEHEMEKAFAFQASQDKVCSICMEVILEKASASERRFGILSNCNHTYCLSCIRQWRCAKQFENPIIKSCPECRVISEFVIPSVYWVEDQNKKNELIEAFKQGMGKKACKYFEQGKGTCPFGSKCLYRHAYPDGRLAEPEKPRKQLSSQGTVRFFNSVRLWDFIENRESRHVPNNEDVDMTELGDLFMHLSGVESSEP

Specifications

Antigen makorin ring finger protein 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human MKRN2 protein.
Formulation PBS with no preservative; pH 7.4
Gene MKRN2
Gene Accession No. NM_014160.3
Gene Alias HSPC070/RNF62
Gene Symbols MKRN2
Host Species Rabbit
Immunogen MKRN2 (NP_054879.3, 1 a.a. ∼ 416 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23609
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.