missing translation for 'onlineSavingsMsg'
Learn More

Mena Mouse, Unlabeled, Clone: 21, BD

Mouse Monoclonal Antibody

332.00€

Specifications

Antigen Mena
Clone 21
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15865589
View Documents
BD Biosciences
610693
150 μg
332.00€
1 set
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Abl and Disabled (Dab) are essential for correct axonal connections during Drosophila development. Mutations in Enabled (Ena) rescue the genetic defects caused by mutated Abl and Dab. Mena (Mammalian Ena) is a protein with significant homology to Eva, VASP, and WASP proteins. The murine Mena gene predicts a protein of 541 amino acids. However, two additional exons that could introduce 246 amino acids onto the C-terminal region have been identified. Homology searches identified two EVH domains (Ena-Vasp homology), one near each terminus of Mena. Antibodies to Mena detect 80 kDa, 88 kDa, and 140 kDa protein with the 80 kDa and 88 kDa proteins being widely expressed and the 140 kDa protein reportedly mainly found in brain tissue. Like VASP, Mena is localized to focal contacts and, when ectopically expressed, induces the formation of F-actin outgrowths in fibroblasts.

Specifications

Specifications

Mena
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgA
Affinity Purified
Mammalian Ena (Enabled)
21
Western Blot
Unconjugated
RUO
Chicken, Human, Mouse, Rat
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.