Learn More
anti-MBD3, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand: Abnova H00053615-PW2.1Set
Includes: Antibody pair for IP: rabbit polyclonal anti-MBD3 (300μL), Antibody pair for WB: mouse purified polyclonal anti-MBD3 (50μg)
Specifications
| MBD3 | |
| Monoclonal/Polyclonal | |
| This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
| MBD3 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-MBD3 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-MBD3 (50 ug) | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| MBD3 | |
| Mouse, Rabbit | |
| RUO | |
| 53615 | |
| Human | |
| Liquid |