missing translation for 'onlineSavingsMsg'
Learn More
Learn More
lymphocyte antigen 6 complex, locus H, Mouse, Clone: 3E10, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant LY6H.
Brand: Abnova H00004062-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: LWCQDCTLTTNSSHCTPKQCQPSDTVCASVRITDPSSSRKDHSVNKMCASSCDFVKRHFFSDYLMGFINSGILKVDVDCCEKDLCNGAAGAGHSPWALAGGLLLSLGPALLWAGPSpecifications
| lymphocyte antigen 6 complex, locus H | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC028894 | |
| LY6H | |
| LY6H (AAH28894, 26 a.a. ∼ 140 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Neurobiology | |
| 4062 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 3E10 | |
| Mouse monoclonal antibody raised against a full length recombinant LY6H. | |
| LY6H | |
| NMLY6 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction