missing translation for 'onlineSavingsMsg'
Learn More

Anti-LTBR antibody [RM2069], Abcam

Product Code. 32090975 Shop All Abcam Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
1 mL
Unit Size:
100µL
1mL
20µL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32090975 1 mL 1mL
32090624 100 μL 100µL
32090773 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 32090975 Supplier Abcam Supplier No. ab3208451mL
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Polyclonal Antibody

Rabbit Recombinant Multiclonal LTBR antibody. Suitable for ICC/IF, Flow Cyt and reacts with Human, Mouse samples.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Ltbr
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence
Cellular Localization Membrane; Single-pass type I membrane protein
Classification Recombinant Polyclonal
Clone RM2069
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene Ltbr
Gene Accession No. P50284
Host Species Rabbit
Immunogen Ltbr
Purification Method Affinity purification
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology & Infectious Disease
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene Ltbr
Target Molecule Ltbr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.