missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-LILRB1, Human, IP-WB Antibody Pair, Abnova™
Description
Includes
Antibody pair for IP: rabbit polyclonal anti-LILRB1 (300μL), Antibody pair for WB: mouse purified polyclonal anti-LILRB1 (50μg)
Specifications
Specifications
| Antigen | LILRB1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | LILRB1 |
| Gene Alias | CD85/CD85J/FLJ37515/ILT2/LIR-1/LIR1/MIR-7/MIR7 |
| Gene Symbols | LILRB1 |
| Host Species | Mouse, Rabbit |
| Quantity | 1 Set |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?