missing translation for 'onlineSavingsMsg'
Learn More

LIM homeobox 4, Mouse, Clone: 4D7, Abnova™

Product Code. 16131077
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16131077 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16131077 Supplier Abnova Supplier No. H00089884M05.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against a partial recombinant LHX4.

This gene encodes a member of a large protein family which contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in control of differentiation and development of the pituitary gland. Mutations in this gene are associated with syndromic short stature and pituitary and hindbrain defects. An alternative splice variant has been described but its biological nature has not been determined. [provided by RefSeq

Sequence: RRAKEKRLKKDAGRHRWGQFYKSVKRSRGSSKQEKESSAEDCGVSDSELSFREDQILSELGHTNRIYGNVGDVTGGQLMNGSFSMDGTGQSYQDLRDGS

Specifications

Antigen LIM homeobox 4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4D7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant LHX4.
Formulation PBS with no preservative; pH 7.4
Gene LHX4
Gene Accession No. NM_033343
Gene Alias Gsh-4/Gsh4
Gene Symbols LHX4
Host Species Mouse
Immunogen LHX4 (NP_203129, 208 a.a. ∼ 306 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 89884
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.