Learn More
anti-LCN2, Human, IP-WB Antibody Pair, Abnova™
Description
Includes
Antibody pair for IP: rabbit polyclonal anti-LCN2 (300μL), Antibody pair for WB: mouse polyclonal anti-LCN2 (50μL)
Specifications
Specifications
| Antigen | LCN2 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | LCN2 |
| Gene Alias | NGAL |
| Gene Symbols | LCN2 |
| Host Species | Mouse, Rabbit |
| Quantity | 1 Set |
| Show More |