Learn More
LAIR-1 Mouse anti-Human, Unlabeled, Clone: DX26, BD
Description
Reacts with leukocyte associated immunoglobulin-like receptor-1 (LAIR-1), a 32kDa transmembrane glycoprotein with a single immunoglobulin-like domain and a cytoplasmic tail containing two immune receptor tyrosine-based inhibitory motifs. It is expressed o
Host Species: Mouse
Clone: DX26
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
Specifications
| Antigen | LAIR-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | DX26 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human NK Cell Clone |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.