missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-KLH, PE-CF594, Clone: X40, BD
Description
Host Species: Mouse
Clone: X40
Isotype: IgG1, κ
Immunogen: Keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Specifications
Specifications
| Antigen | KLH |
| Classification | Monoclonal |
| Clone | X40 |
| Concentration | 0.2mg/mL |
| Conjugate | PE-CF594 |
| Host Species | Mouse |
| Immunogen | Keyhole limpet hemocyanin |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?