Learn More
anti-IL1A Matched Antibody Pair, Ab-1, Abnova
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-IL1A (100μg); Mouse monoclonal anti-IL1A, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. This cytokine is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury, and thus induces apoptosis. This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. It has been suggested that the polymorphism of these genes is associated with rheumatoid arthritis and Alzheimer's disease.
ELISA
Specifications
Specifications
| Antigen | IL1A |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IL1A. |
| Gene | IL1A |
| Gene Alias | IL-1A/IL1/IL1-ALPHA/IL1F1 |
| Gene Symbols | IL1A |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.