missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-7 Rat anti-Human, Unlabeled, Clone: BVD10-40F6, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554493
This item is not returnable.
View return policy
Description
The BVD10-40F6 antibody reacts with human interleukin-7 (IL-7). The immunogen used to generate the BVD10-40F6 hybridoma was recombinant human IL-7. This is a neutralizing antibody.
Specifications
| IL-7 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human IL-7 | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
| ELISA | |
| BVD10-40F6 | |
| Unconjugated | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction