Learn More
IL-5 Rat anti-Mouse/Human, Unlabeled, Clone: TRFK5, BD
Rat Monoclonal Antibody
308.40€
Specifications
| Antigen | IL-5 |
|---|---|
| Clone | TRFK5 |
| Concentration | 0.5mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
Description
The TRFK5 antibody reacts with mouse interleukin-5 (IL-5) and has been reported to cross-react with human IL-5. The TRFK5 antibody has been reported to cross react with IL-5 from rhesus monkey. This is a neutralizing antibody.
Host Species: Rat
Clone: TRFK5
Isotype: IgG1 κ
Species Reactivity: Mouse, Human
Immunogen: Mouse Semi-Purified T-Cell Clone Supernatant
Functional Assay, Neutralization
Specifications
| IL-5 | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Immunology | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
| TRFK5 | |
| ELISA | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Mouse Semi-Purified T-Cell Clone Supernatant | |
| Primary | |
| Store undiluted at 4°C |
For Research Use Only.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.