missing translation for 'onlineSavingsMsg'
Learn More

IL-5 Rat anti-Human, Unlabeled, Clone: JES1-39D10, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 554488

Product Code. 15887728

  • ONLINE EXCLUSIVE PRICE
    382.00€
    381.74€ / Each
    Save 0.26€
    0.07% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The JES1-39D10 antibody reacts with human interleukin-5 (IL-5). The immunogen used to generate the JES1-39D10 hybridoma was COS-expressed recombinant human IL-5. This is a neutralizing antibody.

Host Species: Rat
Clone: JES1-39D10
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Recombinant human IL-5

ELISA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-5
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Recombinant human IL-5
0.5 mg
Immunology
Human
IgG2a
ELISA
JES1-39D10
Unconjugated
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.