Learn More
IL-3 Rat anti-Human, Biotin, Clone: BVD3-1F9, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554674
Description
The BVD3-1F9 antibody reacts with human interleukin-3 (IL-3). The immunogen used to generate the BVD3-1F9 hybridoma was recombinant human IL-3. This is a weakly neutralizing antibody.
Host Species: Rat
Clone: BVD3-1F9
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant Human IL-3
ELISA
Specifications
| IL-3 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human IL-3 | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG1 |
| ELISA | |
| BVD3-1F9 | |
| Biotin | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.