Learn More
anti-IgM, κ, R-PE, Clone: G155-228, BD
Description
The G155-228 antibody is specific for the hapten trinitrophenol (TNP), a hapten not expressed on non-human primate cells or non-human primate cell lines. In the absence of specific binding, this immunoglobulin may bind non-specifically to Fc receptors. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | G155-228 |
| Conjugate | R-PE |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Content And Storage | Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
| Isotype | IgM κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.