missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2a κ, Mouse, Alexa Fluor™ 488, Clone: MOPC-173, BD

Product Code. 15899178
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15899178 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15899178 Supplier BD Biosciences Supplier No. 558055
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The MOPC-173 antibody has unknown specificity. The transplantable plasmacytoma MOPC-173 was induced by intraperitoneal injection of mineral oils into BALB/c mice. In the absence of antigen-specific binding, this immunoglobulin may bind non-specifically to Fc receptors.

Host Species: Mouse
Clone: MOPC-173
Isotype: IgG2a κ

Flow Cytometry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry, Western Blot
Clone MOPC-173
Conjugate Alexa Fluor 488
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Content And Storage Undiluted at 4°C
Isotype IgG2a κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.