Learn More
anti-IgG1 , V450, Clone: X40, BD
Description
Flow Cytometry
Specifications
Specifications
| Antigen | IgG1 |
| Classification | Monoclonal |
| Clone | X40 |
| Conjugate | Violet Fluor 450 (V450) |
| Host Species | Mouse |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Primary |