Learn More
anti-IgG1 κ, Mouse , Pacific Blue™, Clone: MOPC-21, BD
Brand: BD Biosciences 558120
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry, Isotype Control
Specifications
| Flow Cytometry | |
| 0.2mg/mL | |
| Mouse | |
| 0.1 mg | |
| Undiluted at 4°C |
| MOPC-21 | |
| Pacific Blue | |
| Affinity Purified | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.