Learn More
anti-IgG1, κ, Mouse, APC-Cy™7, Clone: MOPC-21, BD
Brand: BD Biosciences 557873
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Flow Cytometry, Isotype Control
Specifications
| Flow Cytometry | |
| APC-Cyanine7 | |
| Affinity Purified | |
| Undiluted at 4°C |
| MOPC-21 | |
| Mouse | |
| 100 Tests | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.