missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG1 κ, Mouse , Alexa Fluor™ 488, Clone: MOPC-21, BD

Product Code. 12890460
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12890460 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12890460 Supplier BD Biosciences Supplier No. 557702
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Clone MOPC-21
Conjugate Alexa Fluor 488
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Content And Storage Undiluted at 4°C
Isotype IgG1 κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.