missing translation for 'onlineSavingsMsg'
Learn More

anti-IgA, κ, Mouse, APC, Clone: M18-254, BD

Product Code. 15851679
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15851679 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15851679 Supplier BD Biosciences Supplier No. 562140

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Clone M18-254
Conjugate APC
Purification Method Affinity Purified
Quantity 0.1 mg
Content And Storage 4°C
Isotype IgA κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.