missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-γ Mouse anti-Human, PE-Cy7, Clone: 4S.B3, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560741
This item is not returnable.
View return policy
Description
The 4S.B3 antibody reacts with human interferon-γ (IFN-γ). The immunogen used to generate this hybridoma was partially purified human IFN-γ obtained from supernatants of human PBMC stimulated with Staphylococcus aureus.
Specifications
| IFN-γ | |
| Monoclonal | |
| PE-Cyanine7 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Baboon, Cynomolgus, Rhesus Monkey | |
| IgG1 κ |
| Flow Cytometry | |
| 4S.B3 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human IFN-γ from supernatants of S. aureus-stimulated PBMC | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction