missing translation for 'onlineSavingsMsg'
Learn More

Anti-IDUA antibody antibody [EPR26926-251], Abcam

Product Code. 32169111 Shop All Abcam Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
1 mL
Unit Size:
100µL
1mL
20µL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32169111 1 mL 1mL
32170753 20 μL 20µL
32167017 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 32169111 Supplier Abcam Supplier No. ab3237011mL
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Rabbit Recombinant Monoclonal IDUA antibody. Suitable for ICC/IF, IHC-P, WB and reacts with Human, Mouse, Rat samples.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen IDUA
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry (FFPE), Western Blot
Cellular Localization Lysosome
Classification Recombinant Monoclonal
Clone EPR26926-251
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene IDUA
Gene Accession No. P35475
Host Species Rabbit
Immunogen IDUA
Purification Method Affinity purification
Quantity 1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Conditional Ambient
Target Gene IDUA
Target Molecule IDUA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.