missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD73 (AD2), Proteintech
Mouse Monoclonal Antibody
100.00€ - 270.00€
Specifications
| Antigen | CD73 |
|---|---|
| Clone | AD2 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235706
|
Proteintech
65162-1-IG-100UG |
100 μg |
100.65€ 100.00€ / 100µg Save 0.65€ 0.65% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237930
|
Proteintech
65162-1-IG-500UG |
500 μg |
271.75€ 270.00€ / 500µg Save 1.75€ 0.64% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD73 antibody from Proteintech is a mouse monoclonal antibody generated with pre-b leukemia cell line of human CD73. This antibody recognizes human antigen. The CD73 antibody has been validated for the following applications: FC analysis.Specifications
| CD73 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 4907 | |
| Pre-B leukemia cell line | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
| AD2 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD73; E5NT; NT; NT5; NTE; eN; eNT | |
| IgG1 κ | |
| Protein A purification | |
| Flow Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title