missing translation for 'onlineSavingsMsg'
Learn More

Anti-Human CD5 (L17F12), Proteintech

Mouse Monoclonal Antibody

50.00€ - 152.00€

Specifications

Antigen CD5
Clone L17F12
Concentration 500 μg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234440
View Documents
Proteintech
65200-1-IG-100UG
100 μg
ONLINE EXCLUSIVE PRICE
52.00€
50.00€ / 100µg
Save 2.00€
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30237431
View Documents
Proteintech
65200-1-IG-500UG
500 μg
ONLINE EXCLUSIVE PRICE
161.00€
152.00€ / 500µg
Save 9.00€
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The CD5 antibody from Proteintech is a mouse monoclonal antibody generated with human t-cell acute lymphoblastic leukemia (t-all) cells of human CD5. This antibody recognizes human antigen. The CD5 antibody has been validated for the following applications: FC analysis.
Specifications

Specifications

CD5
500 μg/mL
Monoclonal
Liquid
RUO
921
Human T-cell acute lymphoblastic leukemia (T-ALL) cells
Primary
Store at 2-8°C. Stable for one year after shipment.
L17F12
Flow Cytometry
Unconjugated
Mouse
Human
LEU1; T1
IgG2a κ
Protein A purification
Flow Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.