missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Human CD196/CCR6; Clone: 11A9; BD
Description
Host Species: Mouse
Clone: 11A9
Isotype: IgG1, κ
Cross Reactivity Species: Hu
Applications:
Flow Cytometry
Specifications
Specifications
| Antigen | Human CD196/CCR6 |
| Classification | Monoclonal |
| Clone | 11A9 |
| Conjugate | Brilliant Violet 605 |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Human |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?