Learn More
Anti-Human CD18 (TS1/18), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65190-1-IG-100UG
This item is not returnable.
View return policy
Description
The CD18 antibody from Proteintech is a mouse monoclonal antibody generated with human beta 2 integrin of human CD18. This antibody recognizes human antigen. The CD18 antibody has been validated for the following applications: FC analysis.Specifications
| CD18 | |
| Monoclonal | |
| 500 μg/mL | |
| CD18; LAD; LCAMB; LFA-1; MAC-1; MF17; MFI7 | |
| Human Beta 2 Integrin | |
| 100 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| TS1/18 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3689 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |