Learn More
Anti-Human CD107a / LAMP1 (H4A3), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65051-1-IG-100UG
This item is not returnable.
View return policy
Description
The CD107a / LAMP1 antibody from Proteintech is a mouse monoclonal antibody generated with human adherent peripheral blood cells of human CD107a / LAMP1. This antibody recognizes human antigen. The CD107a / LAMP1 antibody has been validated for the following applications: WB, IF/ICC, FC (Intra) analysis.Specifications
| CD107a / LAMP1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| CD107a; LAMPA; LGP120 | |
| Human adherent peripheral blood cells | |
| 100 μg | |
| Primary | |
| Human | |
| 100-120 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| H4A3 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3916 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Flow Antibody | |
| IgG1 κ |