Learn More
Anti-Human CD107a / LAMP1 (H4A3), Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65051-1-IG-100UG
 View more versions of this product
Product Code. 30235113
  • ONLINE EXCLUSIVE PRICE
    59.00€
    55.00€ / 100µg
    Save 4.00€
    6% Off
Description
The CD107a / LAMP1 antibody from Proteintech is a mouse monoclonal antibody generated with human adherent peripheral blood cells of human CD107a / LAMP1. This antibody recognizes human antigen. The CD107a / LAMP1 antibody has been validated for the following applications: WB, IF/ICC, FC (Intra) analysis.
Specifications
Specifications
CD107a / LAMP1
Monoclonal
0.5 mg/mL
CD107a; LAMPA; LGP120
Human adherent peripheral blood cells
100 μg
Primary
Human
100-120 kDa
Liquid
Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining)
H4A3
Unconjugated
Mouse
Protein G purification
RUO
3916
Store at 2-8°C. Stable for one year after shipment.
Flow Antibody
IgG1 κ