missing translation for 'onlineSavingsMsg'
Learn More

homeobox D11, Mouse, Clone: 5G4, Abnova™

Product Code. 16169574
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16169574 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16169574 Supplier Abnova Supplier No. H00003237M03.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against a partial recombinant HOXD11.

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located in a cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. The product of the mouse Hoxd11 gene plays a role in forelimb morphogenesis. [provided by RefSeq]

Sequence: MNDFDECGQSAASMYLPGCAYYVAPSDFASKPSFLSQPSSCQMTFPYSSNLAPHVQPVREVAFRDYGLERAKWPYR

Specifications

Antigen homeobox D11
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5G4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HOXD11.
Formulation PBS with no preservative; pH 7.4
Gene HOXD11
Gene Accession No. NM_021192
Gene Alias HOX4/HOX4F
Gene Symbols HOXD11
Host Species Mouse
Immunogen HOXD11 (NP_067015, 1 a.a. ∼ 76 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3237
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.