missing translation for 'onlineSavingsMsg'
Learn More

Anti-HMGA2 antibody [RM1215], Abcam

Product Code. 32096111 Shop All Abcam Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
1 mL
Unit Size:
100µL
1mL
20µL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32096111 1 mL 1mL
32090220 20 μL 20µL
32091440 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 32096111 Supplier Abcam Supplier No. ab3208491mL
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Polyclonal Antibody

Rabbit Recombinant Multiclonal HMGA2 antibody. Suitable for WB, IP, ICC/IF and reacts with Human samples.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen HMGA2
Applications Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot
Cellular Localization Nucleus
Classification Recombinant Polyclonal
Clone RM1215
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene HMGA2
Gene Accession No. P52926
Host Species Rabbit
Immunogen HMGA2
Purification Method Affinity purification
Quantity 1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene HMGA2
Target Molecule HMGA2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.