Learn More
HDAC3, Human, IP-WB Antibody Pair (PW1), Abnova™
Product Code. 16050949
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16050949 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16050949 Supplier Abnova Supplier No. H00008841PW1.1Set
 Restricted Item
Estimated Shipment: 06-10-2026
Includes
Antibody pair for IP: rabbit polyclonal anti-HDAC3 (300μL), Antibody pair for WB: mouse polyclonal anti-HDAC3 (50μL)
For immunoprecipitation and to detect the precipitated protein in western blot.
Specifications
Antigen HDAC3
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene HDAC3
Gene Alias HD3/RPD3/RPD3-2
Gene Symbols HDAC3
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8841
Includes Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-HDAC3 (300 ul) 2. Antibody pair for WB: mouse polyclonal anti-HDAC3 (50 ul)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid