missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-HADH Matched Antibody Pair, Abnova™
Brand: Abnova H00003033-AP51.1Set
Includes: Capture antibody: mouse monoclonal anti-HADH (100μg); Detection antibody: rabbit purified polyclonal anti-HADH (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human HADH
Host Species: Mouse, Rabbit
Species Reactivity: Hu
ELISA Pair (Transfected Lysate)
Specifications
| HADH | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human HADH. | |
| HAD/HADH1/HADHSC/HHF4/M/SCHAD/MGC8392/SCHAD | |
| Mouse, Rabbit | |
| RUO | |
| 3033 | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| HADH | |
| HADH | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-HADH (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-HADH (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction